Merck
  • [A new type of iridium (III) phenylpyrazole complexes: synthesis, photophysical characterization].
[A new type of iridium (III) phenylpyrazole complexes: synthesis, photophysical characterization].
Guang pu xue yu guang pu fen xi = Guang pu (2010-11-26)
Jian-ning Yu, Yu-ying Hao, Xiao-xia Guo, Li-ping Lei, Hui-xia Xu, Bing-she Xu
ABSTRACT
New heteroleptic iridium(III) complexes (ppz)2Ir(LX), which consist of two cyclometalated ligands ppz(1-phenylpyrazole) together with an ancillary ligand LX (LX= 2-(2'-hydroxylphenyl)benzothiazole (BTZ), 2-(3'-methyl-2'-hydroxylphenyl) benzothiazole (3-MeBTZ), 2-(4'-methyl-2'-hydroxylphenyl) benzothiazole (4-MeBTZ) and 2-(4'-Trifluoromethyl-2'hydroxylphenyl) benzothiazole (4-tfmBTZ)), were synthesized and characterized. The molecular structures and photophysical properties were characterized and analyzed comparatively. The results show that the four complexes have basically similar UV-Vis absorption spectra, fluorescence excitation and emission spectra. Their maximum emission peaks are located at 583-615 nm, and accompanied by a lower intensity emission band around 400 nm. The weak emissions around 400 nm are ascribed to the radi ation transition of single state excition from ancillary ligand BTZ perturbed by metallic ion, and light emission around long-wave-length to the radiation transition of 3MLCT of Ir(BTZ) fragment. While the triplet state 3 MLCT of Ir(ppz)2 fragment might be quenched at room temperature. For all complexes, the excitations with maximum efficiency are located at 250-310 nm, which indicates that main contributor to light emitting is ligand-centered absorption (1pi-pi*) of ppz and BTZ rather than 3MLCT transitions, and thus provides a striking evidence that there is intersystem crossing from 1pi-pi* state to 3MLCT state in these complexes. Compared with Ir(ppz)3, these complexes not only have stronger phosphorescence at room temperature but also their emission color can be tuned by modifying ancillary ligand.
Sigma-Aldrich
1-Phenylpyrazole, 97%