Merck
Get up to 22% off for Pi Day until 3/26.Save Now
Total synthesis of (+)-7-deoxypancratistatin from furan.
Organic letters (2000-11-14)
J L Aceña, O Arjona, M L León, J Plumet
ABSTRACT
A new total synthesis of (+)-7-deoxypancratistatin 1 has been accomplished in 19 steps (8% overall yield) from two readly available compounds, furan and trans-1,2-bis(phenylsulfonyl)ethylene.
Sigma-Aldrich
trans-1,2-Bis(phenylsulfonyl)ethylene, 98%