Merck
  • Helical molecular programming: folding of oligopyridine-dicarboxamides into molecular single helices.
Helical molecular programming: folding of oligopyridine-dicarboxamides into molecular single helices.
Chemistry (Weinheim an der Bergstrasse, Germany) (2001-08-07)
V Berl, I Huc, R G Khoury, J M Lehn
ABSTRACT
Molecular strands composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units have been designed to self-organize into single stranded helical structures upon forming intramolecular hydrogen bonds. Pentameric strands 11, 12, and 14, heptameric strands 1 and 20, and undecameric strand 15 have been synthesized using stepwise convergent strategies. Single helical conformations have been characterized in the solid state by single crystal X-ray diffraction analysis for four of these compounds. Helices from pentameric strands 12 and 14 extend over one turn, and helices from heptameric 20 and undecameric 15 species extend to one and a half and two and a half turns, respectively. Intramolecular hydrogen bonds are responsible for the strong bending of the strands. 1H NMR shifts both in polar and nonpolar organic solvents indicate intramolecular overlap between the peripheral aromatic groups. Thus, helical conformations also predominate in solution. Molecular stochastic dynamic simulations of strand folding starting from a high energy extended linear conformer show a rapid (600 ps at 300 K) conversion into a stable helical conformation.
Sigma-Aldrich
Decanoyl chloride, 98%