- Effects of inhibitors of hydroxymethylglutaryl coenzyme A reductase on coenzyme Q and dolichol biosynthesis.
Effects of inhibitors of hydroxymethylglutaryl coenzyme A reductase on coenzyme Q and dolichol biosynthesis.
Inhibitors of hydroxymethylglutaryl coenzyme A reductase are used clinically to decrease blood levels of low-density lipoprotein cholesterol in hypercholesterolemic patients. However, little is known about the possible effects of these inhibitors on dolichol and cholesterol synthesis. Oral administration of mevinolin to rats was found here to decrease dolichol, dolichyl-P and coenzyme Q levels in the heart and skeletal muscle and to increase the hepatic dolichol level while decreasing the coenzyme Q content in this same organ. The amounts of dolichyl-P decreased in heart and muscle and increased in brain. Intraperitoneal administration also affected the levels of these lipids. The concentrations of blood lipids were not modified in the same manner as tissue lipids. Analysis of individual enzyme activities and of incorporation of [3H]acetate into various lipids of liver and brain slices demonstrated that both up- and down-regulation of different proteins occur in various tissues, resulting in modifications in lipid synthesis. Hypercholesterolemic patients were found to have high blood coenzyme Q levels, which are decreased upon pravastatin treatment, although they are still above control values. It appears that these HMG-coenzyme A reductase inhibitors do not selectively lower cholesterol levels, but that they also modify the dolichol and coenzyme Q content and synthesis both in the liver and various other tissues.