Merck
  • Photodegradation of protoporphyrin-dimethylester in solution and in organized environments.
Photodegradation of protoporphyrin-dimethylester in solution and in organized environments.
International journal of radiation biology (1993-11-01)
J M Wessels, R Sroka, P Heil, H K Seidlitz
ABSTRACT
The degradation of sensitizers used in photodynamic therapy (PDT) involves photooxidation either by molecular oxygen or by oxygen intermediates which leads to hydroxyaldehyde and formyl products or to ring opening. Our investigations focused on the spectroscopic changes which protoporphyrin-dimethylester (PP) exhibits upon irradiation. As the microenvironment strongly influences the effects, we used an aprotic organic solvent, L-alpha-phosphatidylcholine dioleoyl (DOPC) liposomes and isogenic fibrosarcoma cells (SSKII) as carriers for PP. Hydroxyaldehyde product isomers develop a new absorption band centred around 670 nm and a new emission band at 676 nm. These characteristics can be used to discriminate them from formyl products and intact PP. In organic solvents, the formation of the hydroxyaldehyde products dominates. In DOPC liposomes and cells, the hydroxyaldehyde yield drops and photooxidation results in attack of the macrocycle. Time-resolved fluorescence spectroscopy of monomeric PP in an organic solvent gives a monoexponential decay time tau of 10.1 +/- 1.3 ns. Upon irradiation a second component with a decay time of 4.9 +/- 0.6 ns, resulting from the hydroxyaldehyde product, was detected. In liposomes and cells the monomeric decay time was significantly longer (15 ns) due to the altered microenvironment. Additionally, we observed in liposomes and in cells a small contribution of a short component (1 ns) which is attributed to an aggregated sensitizer species. In irradiated cells the aggregated fraction doubles, indicating a change in the microenvironment caused by the photodynamic action of the sensitizer.
Sigma-Aldrich
Protoporphyrin IX dimethyl ester, ~90% (HPLC)