Merck
  • Effect of 5-methoxytryptophan and 5-methoxytryptamine on the reproductive system of the male golden hamster.
Effect of 5-methoxytryptophan and 5-methoxytryptamine on the reproductive system of the male golden hamster.
Journal of neural transmission (1981-01-01)
P Pévet, C Haldar-Misra, T Ocal
ABSTRACT
S.c. injection of 25 microgram of methoxytryptamine (5-MT) in oil into adult male hamsters given between 4.30 and 5 p.m. (light on from 5 a.m. to 7 p.m.: 14 L/10 D) for 51 consecutive days caused involution of the testes. Similar injections of 5-MT given between 8.30 and 9.30 a.m. completely failed to cause regression of the gonads. Under the same conditions, 5-methoxytryptophan (5-MTP) did not induce gonadal atrophy, neither in the morning nor in the late afternoon. These results indicate that, like malatonin, 5-methoxytryptamine is implicated in the control of the reproductive function.
Sigma-Aldrich
5-Methoxy-DL-tryptophan