Merck
  • [Disease of the fins of Rhamdia sapo. Isolation of the etiological agents and experimental infection].
[Disease of the fins of Rhamdia sapo. Isolation of the etiological agents and experimental infection].
Revista Argentina de microbiologia (1988-01-01)
N M Angelini, G N Seigneur
ABSTRACT
During intensive culture of South-American catfish, Rhamdia sapo fingerlings an epizootic similar to fin rot was developed. Aeromonas hydrophila and Pseudomonas aeruginosa were isolated and they were treated and controlled with daily short dip, 1 ppm Furanace. The disease was experimentally reproduced and we concluded that it is result of interaction of unfavorable environment conditions such as: low dissolved oxygen, increase of solids in suspension; host condition and the presence of Aeromonas hydrophila and Pseudomonas aeruginosa.