Merck
  • The D₁ dopamine receptor agonist, SKF83959, attenuates hydrogen peroxide-induced injury in RGC-5 cells involving the extracellular signal-regulated kinase/p38 pathways.
The D₁ dopamine receptor agonist, SKF83959, attenuates hydrogen peroxide-induced injury in RGC-5 cells involving the extracellular signal-regulated kinase/p38 pathways.
Molecular vision (2012-12-13)
Guang-Yu Li, Ting Li, Bin Fan, Yong-Chen Zheng, Tong-Hui Ma
ABSTRACT
Oxidative stress is widely implicated in the death of retinal ganglion cells associated with various optic neuropathies. Agonists of the dopamine D(1) receptor have recently been found to be potentially neuroprotective against oxidative stress-induced injury. The goal of this study was to investigate whether SKF83959, a next-generation high-affinity D(1) receptor agonist, could protect retinal ganglion cell 5 (RGC-5) cells from H(2)O(2)-induced damage and the molecular mechanism involved. We examined expression of the D(1) receptor in RGC-5 cells with reverse-transcription-PCR and immunoblotting and assessed neuroprotection using propidium iodide staining and the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay. In addition, we monitored the activation and involvement of members of mitogen-activated protein kinase family, extracellular signal-regulated kinase (ERK), p38 and c-Jun NH(2)-terminal kinase, with western blot and specific inhibitors. We found that the D(1) receptor was expressed in RGC-5 cells, but the sequence analysis suggested this cell line is from mouse and not rat origin. SKF83959 exhibited a remarkable neuroprotective effect on H(2)O(2)-damaged RGC-5 cells, which was blocked by the specific D(1) receptor antagonist, SCH23390. ERK and p38 were activated by SKF83959, and pretreatment with their inhibitors U0126 and SB203580, respectively, significantly blunted the SKF83959-induced cytoprotection. However, the specific c-Jun NH(2)-terminal kinase inhibitor, SP600125, had no effect on the SKF83959-induced protection. We conclude that SKF83959 attenuates hydrogen peroxide-induced injury in RGC-5 cells via a mechanism involving activation of the ERK and p38 pathways and the D(1) receptor is a potential molecular target for developing neuroprotective drugs.
Sigma-Aldrich
SKF-83959 hydrobromide, ≥98% (HPLC), solid