Merck
  • Nephrotoxicity of hexachloro-1:3-butadiene in the male Hanover Wistar rat; correlation of minimal histopathological changes with biomarkers of renal injury.
Nephrotoxicity of hexachloro-1:3-butadiene in the male Hanover Wistar rat; correlation of minimal histopathological changes with biomarkers of renal injury.
Journal of applied toxicology : JAT (2011-09-10)
Aubrey Swain, John Turton, Cheryl Scudamore, David Maguire, Ines Pereira, Sofia Freitas, Rosemary Smyth, Michael Munday, Clare Stamp, Mitul Gandhi, Surjit Sondh, Holly Ashall, Ian Francis, Jennifer Woodfine, John Bowles, Malcolm York
ABSTRACT
Hexachloro-1:3-butadiene (HCBD) causes damage specifically to the renal proximal tubule in rats. In the present study, injury to the nephron of male Hanover Wistar rats was characterized at 24 h following dosing with HCBD in the range 5-90 mg kg⁻¹ to determine the most sensitive biomarkers of damage, that is, the biomarkers demonstrating significant changes at the lowest dose of HCBD, using a range of measurements in serum and urine, renal histopathology, and renal and hepatic gene expression. Histologically, kidney degeneration was noted at doses as low as 10 mg kg⁻¹ HCBD. Significant changes in the hepatic and renal gene expression categories of xenobiotic metabolism and oxidative stress were observed at 5 mg kg⁻¹ HCBD, and in the kidney alone, evidence of inflammation at 90 mg kg⁻¹ HCBD. Increases in the urinary excretion of α-glutathione S-transferase (α-GST) and kidney injury molecule-1 (KIM-1) were seen at 10 mg kg⁻¹ HCBD, and increases in urinary excretion of albumin and total protein were evident at 15 mg kg⁻¹ HCBD. The most sensitive, noninvasive biomarkers of HCBD-induced renal toxicity in Hanover Wistar rats were urinary α-GST and KIM-1. Urinary albumin measurement is also recommended as, although it is not the most sensitive biomarker, together with α-GST, albumin showed the largest relative increase of all the biomarkers investigated, and the protein is easily measured.
Sigma-Aldrich
Hexachloro-1,3-butadiene, 96%
Supelco
Hexachloro-1,3-butadiene, analytical standard