Merck
Enantiospecific wetting.
Journal of the American Chemical Society (2010-12-04)
Michael Rapp, William A Ducker
ABSTRACT
Experiments have shown that enantiospecificity can be important in wetting. Measurements of droplet contact angles can be used to estimate the energy of enantiomeric discrimination.