Merck
  • Toward homogeneous erythropoietin: fine tuning of the C-terminal acyl donor in the chemical synthesis of the Cys29-Gly77 glycopeptide domain.
Toward homogeneous erythropoietin: fine tuning of the C-terminal acyl donor in the chemical synthesis of the Cys29-Gly77 glycopeptide domain.
Journal of the American Chemical Society (2009-04-22)
Yu Yuan, Jin Chen, Qian Wan, Zhongping Tan, Gong Chen, Cindy Kan, Samuel J Danishefsky
ABSTRACT
Described herein is the chemical synthesis of the Cys(29)-Gly(77) glycopeptide domain (22) of erythropoietin. Our initial ligation strategy targeted a C --> N termini condensation between glycopeptide 3 and peptide 4. However, the reaction was hindered by the "unattainable" reactivity, mismatched polarity, and severe aggregation of the (glyco)peptide substrates. In contrast, by tuning the C-terminal acyl donor and using smaller peptide fragments, the Cys(29)-Gly(77) glycopeptide domain of erythropoietin was prepared through unconventional N --> C termini condensation reactions. The use of a p-cyanonitrophenyl ester and the development of a masked thiophenyl ester as acyl donors enabled us to promptly access glycopeptides bearing complex carbohydrates and offer potential synthetic applications beyond our current work.
Sigma-Aldrich
S-Phenyl thioacetate, 98%