Merck
  • Adenosine (A2) antagonist inhibits induction of long-term potentiation of evoked synaptic potentials but not of the population spike in hippocampal CA1 neurons.
Adenosine (A2) antagonist inhibits induction of long-term potentiation of evoked synaptic potentials but not of the population spike in hippocampal CA1 neurons.
Biochemical and biophysical research communications (1991-12-31)
Y Sekino, K Ito, H Miyakawa, H Kato, Y Kuroda
ABSTRACT
The effects of adenosine A2 receptor antagonist (CP-66713) on long-term potentiation were studied using guinea pig hippocampal slices in a perfusion system. Tetanic stimulation of Schaffer collateral input which was applied during perfusion of CP-66713 (10 microM), did not induce long-term potentiation but rather long-term depression of evoked synaptic potentials (field EPSP), but induced long-term potentiation of the population spike in CA1 neurons. Thus, adenosine derivatives which accumulate in the synaptic cleft during the tetanic stimulation may be involved in induction of the long-term potentiation via A2 receptors at the synapse. The clear discrimination between long-term depression of the field EPSP and long-term potentiation of the population spike suggests EPSP-spike potentiation at the postsynaptic sites.
Sigma-Aldrich
CP-66713, ≥98% (HPLC)