Merck
  • Determination of metolazone in human blood by liquid chromatography with electrospray ionization tandem mass spectrometry.
Determination of metolazone in human blood by liquid chromatography with electrospray ionization tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-08-16)
Guangli Wei, Shuhua Xiao, Changxiao Liu
ABSTRACT
A rapid, sensitive and accurate liquid chromatographic-tandem mass spectrometric method is described for the determination of metolazone in human blood. Metolazone was extracted from blood using ethyl acetate and separated on a C18 column interfaced with a triple quadrupole tandem mass spectrometer. The mobile phase consisting of a mixture of acetonitrile, 10 mmol/l ammonium acetate and formic acid (60:40:0.1, v/v/v) was delivered at a flow rate of 0.5 ml/min. Electrospray ionization (ESI) source was operated in positive ion mode. Selected reaction monitoring (SRM) mode using the transitions of m/z 366-->m/z 259 and m/z 321-->m/z 275 were used to quantify metolazone and the lorazepam (internal standard), respectively. The linearity was obtained over the concentration range of 0.5-500 ng/ml for metolazone and the lower limit of quantitation (LLOQ) was 0.5 ng/ml. For each level of QC samples, inter- and intra-run precision was less than 8.07 and 3.56% (relative standard deviation (RSD)), respectively, and the bias was within +/-4.0%. This method was successfully applied to the pharmacokinetic study of metolazone formulation after oral administration to humans.
Sigma-Aldrich
Metolazone, ≥98% (HPLC), solid