Merck
  • Fully stereocontrolled syntheses of 3-oxacarbacyclin and carbacyclin by the conjugate addition-azoalkene-asymmetric olefination strategy.
Fully stereocontrolled syntheses of 3-oxacarbacyclin and carbacyclin by the conjugate addition-azoalkene-asymmetric olefination strategy.
The Journal of organic chemistry (2006-06-06)
Mikhail Kim, Hans-Joachim Gais
ABSTRACT
A fully stereocontrolled synthesis of 3-oxacarbacyclin (3) and a formal synthesis of carbacyclin (2) are described. The syntheses are based on the conjugate addition-azoalkene-asymmetric olefination strategy. Its key features are (1) the stereoselective establishment of the complete omega-side chain of 2 and 3 through conjugate addition of the enantiopure C13-C20 alkenylcopper derivative 10 to the enantiopure C6-C12 bicyclic azoalkene 9 and (2) the 5E-stereoselective construction of the alpha-side chain through a Horner-Wadsworth-Emmons olefination of the bicyclic ketone 7 with the chiral lithium phosphonoacetate 26 with formation of ester E-27. The allylic alcohol 6 serves at late stage as the joint intermediate in the synthesis of 2 and 3.