- [An experimental study of ocular contusion and its hemodynamic changes].
[An experimental study of ocular contusion and its hemodynamic changes].
[Zhonghua yan ke za zhi] Chinese journal of ophthalmology (1992-07-01)
H L Dou, C Song
PMID1299573
ABSTRACT
The hemodynamic changes in rabbit eyes following contusion were studied with rheoophthalmography (ROG), in which the wave amplitudes were markedly reduced and then recovered in 3 weeks, while in the anisodine-I treated group they recovered in 2 weeks. Contusion of 3 joules disrupted the choroidal circulation and damaged the retina. Anisodine-I improved the vasomotor functions of the choroidal vessels and was effective in treatment of ocular contusion. ROG was a sensitive objective means of estimating the severity of ocular contusion and evaluating the therapeutic effects.