Merck
  • Human urotensin II-induced contraction and arterial smooth muscle cell proliferation are mediated by RhoA and Rho-kinase.
Human urotensin II-induced contraction and arterial smooth muscle cell proliferation are mediated by RhoA and Rho-kinase.
Circulation research (2001-06-09)
V Sauzeau, E Le Mellionnec, J Bertoglio, E Scalbert, P Pacaud, G Loirand
ABSTRACT
The aim of this work was to investigate the coupling of human urotensin II (hU-II) to RhoA activation and regulation of RhoA-dependent functions. The use of the Rho-kinase inhibitor Y-27632 and the development of a membrane-permeant RhoA inhibitor (TAT-C3) allowed us to demonstrate that hU-II induced arterial smooth muscle contraction, actin stress fiber formation, and proliferation through the activation of the small GTPase RhoA and its downstream effector Rho-kinase.
Sigma-Aldrich
Urotensin II human, ≥97% (HPLC), synthetic