Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Degradation of 4-chloro-2-methylphenol by an activated sludge isolate and its taxonomic description.
Degradation of 4-chloro-2-methylphenol by an activated sludge isolate and its taxonomic description.
Biodegradation (1995-06-01)
U Lechner, R Baumbach, D Becker, V Kitunen, G Auling, M Salkinoja-Salonen
ABSTRACT
The Gram-negative strain S1, isolated from activated sludge, metabolized 4-chloro-2-methylphenol by an inducible pathway via a modified ortho-cleavage route as indicated by a transiently secreted intermediate, identified as 2-methyl-4-carboxymethylenebut-2-en-4-olide by gas chromatography/mass spectrometry. Beside 4-chloro-2-methylphenol only 2,4-dichlorophenol and 4-chlorophenol were totally degraded, without an accumulation of intermediates. The chlorinated phenols tested induced activities of 2,4-dichlorophenol hydroxylase and catechol 1,2-dioxygenase type II. Phenol itself appeared to be degraded more efficiently via a separate, inducible ortho-cleavage pathway. The strain was characterized with respect to its physiological and chemotaxonomic properties. The fatty acid profile, the presence of spermidine as main polyamine, and of ubiquinone Q-10 allowed the allocation of the strain into the alpha-2 subclass of the Proteobacteria. Ochrobactrum anthropi was indicated by fatty acid analysis as the most similar organism, however, differences in a number of physiological features (e.g. absence of nitrate reduction) and pattern of soluble proteins distinguished strain S1 from this species.
Product Description
Supelco
4-Chloro-2-methylphenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chloro-2-methylphenol, 97%