Merck
  • 1H-nuclear magnetic resonance of intercalators and rGCA: a potential mutagenicity probe.
1H-nuclear magnetic resonance of intercalators and rGCA: a potential mutagenicity probe.
Chemico-biological interactions (1984-06-01)
J M Coddington, D Alkema, R A Bell, D W Hughes, T Neilson
ABSTRACT
Variable temperature 1H-nuclear magnetic resonance (NMR) has been used to study the interaction of the RNA trimer, GpCpA, with the intercalators ethidium bromide and the acridine derivatives; proflavin, 9-amino-acridine, acridine orange, acridine yellow and acriflavin. The complexes formed were studied at nucleic acid to drug ratios of 1:1 and 5:1, the latter being useful in defining the effects of structural variation in the acridine series and in determining the site of intercalation. All the intercalators greatly stabilized the oligonucleotide duplex, the average melting temperature (Tm) increasing by up to 30 degrees C. Significant changes in individual Tms and chemical shifts were observed for all the GpCpA protons. 9-Amino-acridine and acriflavin did not stabilize the GpCpA duplex as substantially as the other acridine derivatives. It is suggested that this intercalator:GpCpA system, and its associated NMR-derived Tm, is a useful physical probe for potential mutagens.
Sigma-Aldrich
Acridine Yellow G, Dye content 90 %