Merck
  • Translocation of PKC-betaII is mediated via RACK-1 in the neuronal cells following dioxin exposure.
Translocation of PKC-betaII is mediated via RACK-1 in the neuronal cells following dioxin exposure.
Neurotoxicology (2006-06-27)
Hyun-Gyo Lee, Sun-Young Kim, Eun-Jung Choi, Ki-Yeon Park, Jae-Ho Yang
ABSTRACT
2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD) is known to induce neurotoxic effects. However, the mechanism of TCDD-mediated signaling pathways and its possible molecular targets in neurons remains unknown. In this study, we analyzed effects of TCDD on neurofilament subunits, receptor for activated C kinase-1 (RACK-1), and PKC-betaII activity in developing neuronal cells. TCDD induced a significant increase of RACK-1, an adaptor protein for protein kinase C (PKC), in cerebellar granule cells in both dose- and time-dependent manner, indicating that RACK-1 is a sensitive molecular target in neuronal cells for TCDD exposure. TCDD induced a dose-dependent translocation of PKC-betaII from cytosol to membrane fractions. However, when RACK-1 induction was blocked by antisense oligonucleotide or alpha-naphthoflavone, Ah receptor (AhR) inhibitor, the translocation of PKC-betaII was inhibited. Our data suggests that TCDD activates PKC-betaII via RACK-1 in an AhR-dependent manner. This is the first report identifying RACK-1 as a target molecule involved in TCDD-mediated signaling pathways. TCDD exposure also increased the level of neurofilament-H mRNA. These results suggest that identification of target molecules may contribute to improve our understanding of TCDD-mediated signaling pathway and the risk assessment of TCDD-induced neurotoxicities.