Merck
  • ApoG2 as the most potent gossypol derivatives inhibits cell growth and induces apoptosis on gastric cancer cells.
ApoG2 as the most potent gossypol derivatives inhibits cell growth and induces apoptosis on gastric cancer cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2012-12-04)
J Xin, Y H Zhan, L M Xia, H W Zhu, Y Z Nie, J M Liang, J Tian
ABSTRACT
Gastric cancer is one of the most common types of malignancies and proteins from the Bcl-2 family are highly expressed in human gastric cancer. Apogossypolone (ApoG2), the most potent gossypol derivative, has been defined as a novel small-molecule inhibitor of anti-apoptotic Bcl-2 family proteins. However, whether or not it can inhibit the growth and proliferation of gastric cancer cell lines has not been demonstrated to date. Here, we assessed the effects of anti-growth of ApoG2 on gastric cancer cell lines in vitro and explored the possible molecular mechanisms of ApoG2. Using the MTT assay and flow cytometry, we found that ApoG2 has the significant anti-growth effect on MKN28, MKN45 and AGS cell lines in a time- and dose-dependent manner. Compared to (-)-gossypol, MTT assay and flow cytometry results showed that anti-growth effect of ApoG2 is inferior, but the colony formation ability of ApoG2 is superior. Furthermore, western blot results revealed that ApoG2 inhibits the growth and proliferation of gastric cancer cells by down-regulating of Bcl-2 protein expression, up-regulating of Bax and activating of Caspase-3. Taken together, albeit the ApoG2 inferior to (-)-gossypol in many ways on gastric cancer in vitro, our results suggest that ApoG2 could effectively inhibit the growth and proliferation of gastric cancer cell lines through the mitochondrial pathway of apoptosis.
Sigma-Aldrich
(±)-Gossypol from cotton seeds, ≥95% (HPLC)