Merck
  • Extra- and intracellular imaging of human matrix metalloprotease 11 (hMMP-11) with a cell-penetrating FRET substrate.
Extra- and intracellular imaging of human matrix metalloprotease 11 (hMMP-11) with a cell-penetrating FRET substrate.
The Journal of biological chemistry (2012-08-29)
B Sina Meyer, Jörg Rademann
ABSTRACT
Matrix metalloprotease 11 (MMP-11), a protease associated with invasion and aggressiveness of cancerous tissue, was postulated as a prognostic marker for pancreatic, breast, and colon cancer patients. Expression analysis, however, did not reveal localization and regulation of this protease. Thus, cellular tools for the visualization of MMP-11 are highly desirable to monitor presence and activity and to elucidate the functional role of MMP-11. Therefore, fluorescein-Dabcyl-labeled Foerster resonance energy transfer (FRET) substrates were developed. The design focused on enhanced peptide binding to human MMP-11, employing an unusual amino acid for the specificity pocket P1'. The addition of several arginines resulted in a cell-permeable FRET substrate SM-P124 (Ac-GRRRK(Dabcyl)-GGAANC(MeOBn)RMGG-fluorescein). In vitro evaluation of SM-P124 with human MMP-11 showed a 25-fold increase of affinity (k(cat)/K(m) = 9.16 × 10(3) m(-1) s(-1), K(m) = 8 μm) compared with previously published substrates. Incubation of pancreatic adenocarcinoma cell line MIA PaCa-2 and mamma adenocarcinoma cell line MCF-7 with the substrate SM-P124 (5 μm) indicated intra- and extracellular MMP-11 activity. A negative control cell line (Jurkat) showed no fluorescent signal either intra- or extracellularly. Negative control FRET substrate SM-P123 produced only insignificant extracellular fluorescence without any intracellular fluorescence. SM-P124 therefore enabled intra- and extracellular tracking of MMP-11-overexpressing cancers such as pancreatic and breast adenocarcinoma and might contribute to the understanding of the activation pathways leading to MMP-11-mediated invasive processes.
Product Description
Supelco
Butter Yellow, analytical standard