Merck
  • Structure-functional selectivity relationship studies of β-arrestin-biased dopamine D₂ receptor agonists.
Structure-functional selectivity relationship studies of β-arrestin-biased dopamine D₂ receptor agonists.
Journal of medicinal chemistry (2012-08-01)
Xin Chen, Maria F Sassano, Lianyou Zheng, Vincent Setola, Meng Chen, Xu Bai, Stephen V Frye, William C Wetsel, Bryan L Roth, Jian Jin
ABSTRACT
Functionally selective G protein-coupled receptor (GPCR) ligands, which differentially modulate canonical and noncanonical signaling, are extremely useful for elucidating key signal transduction pathways essential for both the therapeutic actions and side effects of drugs. However, few such ligands have been created, and very little purposeful attention has been devoted to studying what we term: "structure-functional selectivity relationships" (SFSR). We recently disclosed the first β-arrestin-biased dopamine D(2) receptor (D(2)R) agonists UNC9975 (44) and UNC9994 (36), which have robust in vivo antipsychotic drug-like activities. Here we report the first comprehensive SFSR studies focused on exploring four regions of the aripiprazole scaffold, which resulted in the discovery of these β-arrestin-biased D(2)R agonists. These studies provide a successful proof-of-concept for how functionally selective ligands can be discovered.