Merck
  • Determination of traces of iron(II) in the presence of iron(III) by the bathophenanthroline method.
Determination of traces of iron(II) in the presence of iron(III) by the bathophenanthroline method.
Talanta (1972-03-01)
T Mizuno
ABSTRACT
Traces of iron(II) (1-30 ppM) in the presence of iron(III) were determined (error <10%) by the bathophenanthroline method. Interference of iron(III) was eliminated by masking with sodium pyrophosphate (2.5-60 mg). The iron(II) complex was extracted with n-butanol, at pH 4.2-4.7.
Product Description
Supelco
Bathophenanthroline, for spectrophotometric det. of Fe in serum, ≥99.0%