Merck
  • Primary hyperoxaluria type III--a model for studying perturbations in glyoxylate metabolism.
Primary hyperoxaluria type III--a model for studying perturbations in glyoxylate metabolism.
Journal of molecular medicine (Berlin, Germany) (2012-06-26)
Ruth Belostotsky, James Jonathon Pitt, Yaacov Frishberg
ABSTRACT
Perturbations in glyoxylate metabolism lead to the accumulation of oxalate and give rise to primary hyperoxalurias, recessive disorders characterized by kidney stone disease. Loss-of-function mutations in HOGA1 (formerly DHDPSL) are responsible for primary hyperoxaluria type III. HOGA1 is a mitochondrial 4-hydroxy-2-oxoglutarate aldolase catalyzing the fourth step in the hydroxyproline pathway. We investigated hydroxyproline metabolites in the urine of patients with primary hyperoxaluria type III using gas chromatography-mass spectroscopy. Significant increases in concentrations of 4-hydroxy-2-oxoglutarate and its precursor and derivative 4-hydroxyglutamate and 2,4-dihydroxyglutarate, respectively, were found in all patients as compared to carriers of the corresponding mutations or healthy controls. Despite a functional block in the conversion of hydroxyproline to glyoxylate--the immediate precursor of oxalate--the production of oxalate increases. To explain this apparent contradiction, we propose a model of glyoxylate compartmentalization in which cellular glyoxylate is normally prevented from contact with the cytosol where it can be oxidized to oxalate. We propose that HOGA1 deficiency results in the accumulation of 4-hydroxy-2-oxoglutarate in the mitochondria and its transport into the cytosol where it is converted to glyoxylate by a different cytosolic aldolase. In human hepatocyte cell lines, we detected a cytosolic 4-hydroxy-2-oxoglutarate aldolase activity not due to HOGA1. These studies provide a diagnostic tool for primary hyperoxaluria type III and shed light on glyoxylate metabolism and the pathogenesis of primary hyperoxalurias.
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O