Merck
  • Extractive spectrophotometric determination of palladium with N,N,N',N'-tetra(2-ethylhexyl)-thiodiglycolamide T(2EH)TDGA.
Extractive spectrophotometric determination of palladium with N,N,N',N'-tetra(2-ethylhexyl)-thiodiglycolamide T(2EH)TDGA.
Talanta (2011-07-06)
R Ruhela, J N Sharma, B S Tomar, R C Hubli, A K Suri
ABSTRACT
A precise, sensitive and selective method for the spectrophotometric determination of palladium (II) using N,N,N',N'-tetra(2-ethylhexyl) thiodiglycolamide T(2EH)TDGA as an extractant is described. Palladium (II) forms yellow colored complex with T(2EH)TDGA which exhibits an absorption maximum at ∼ 300 nm. The colored complex obeys Beer's law in the concentration range 1.0-15.0 μg ml(-1) of palladium with a molar absorptivity of 1.29 × 10(5)M(-1)cm(-1). The effects of various experimental parameters have been studied to establish the optimum conditions for the extraction and determination of palladium. The precision of the method has been evaluated and the relative standard deviation has been found to be less than 0.5%. The method has been successfully applied to the determination of palladium in simulated high level liquid waste (SHLW) solution.
Sigma-Aldrich
Glycolamide, 98%