Merck
  • On the performance of spin diffusion NMR techniques in oriented solids: prospects for resonance assignments and distance measurements from separated local field experiments.
On the performance of spin diffusion NMR techniques in oriented solids: prospects for resonance assignments and distance measurements from separated local field experiments.
The journal of physical chemistry. B (2010-10-13)
Nathaniel J Traaseth, T Gopinath, Gianluigi Veglia
ABSTRACT
NMR spin diffusion experiments have the potential to provide both resonance assignment and internuclear distances for protein structure determination in oriented solid-state NMR. In this paper, we compared the efficiencies of three spin diffusion experiments: proton-driven spin diffusion (PDSD), cross-relaxation-driven spin diffusion (CRDSD), and proton-mediated proton transfer (PMPT). As model systems for oriented proteins, we used single crystals of N-acetyl-L-(15)N-leucine (NAL) and N-acetyl-L-(15)N-valyl-L-(15)N-leucine (NAVL) to probe long and short distances, respectively. We demonstrate that, for short (15)N/(15)N distances such as those found in NAVL (3.3 Å), the PDSD mechanism gives the most intense cross-peaks, while, for longer distances (>6.5 Å), the CRDSD and PMPT experiments are more efficient. The PDSD was highly inefficient for transferring magnetization across distances greater than 6.5 Å (NAL crystal sample), due to small (15)N/(15)N dipolar couplings (<4.5 Hz). Interestingly, the mismatched Hartmann-Hahn condition present in the PMPT experiment gave more intense cross-peaks for lower (1)H and (15)N RF spinlock amplitudes (32 and 17 kHz, respectively) rather than higher values (55 and 50 kHz), suggesting a more complex magnetization transfer mechanism. Numerical simulations are in good agreement with the experimental findings, suggesting a combined PMPT and CRDSD effect. We conclude that, in order to assign SLF spectra and measure short- and long-range distances, the combined use of homonuclear correlation spectra, such as the ones surveyed in this work, are necessary.
Sigma-Aldrich
N-Acetyl-L-leucine, ReagentPlus®, 99%