Merck
  • Unraveling the hydroxypropionaldehyde (HPA) system: an active antimicrobial agent against human pathogens.
Unraveling the hydroxypropionaldehyde (HPA) system: an active antimicrobial agent against human pathogens.
Journal of agricultural and food chemistry (2010-09-18)
Sabine Vollenweider, Stefan Evers, Karin Zurbriggen, Christophe Lacroix
ABSTRACT
The hydroxypropionaldehyde (HPA) system is a natural defense system synthesized by the probiotic bacterium Lactobacillus reuteri. To elucidate which of the molecules composing the HPA system (3-hydroxypropionaldehyde (3-HPA), reuterin (HPA dimer), and HPA hydrate) is responsible for the potent antimicrobial activity in biological systems, a combination of biochemical, genetic, and proteomic assays was used. The HPA system reacts with sulfhydryl-containing compounds such as cysteine and reduced glutathione (GSH) in solution. In situ, GSH knock-out Escherichia coli is significantly more susceptible to HPA-mediated cell death than E. coli wild type; GSH supplementation protects either bacteria from HPA attack. Proteomic analysis of HPA-treated bacteria ( Haemophilus influenzae ) revealed induction of redox- and heat shock-related proteins. A new antimicrobial mechanism of HPA is proposed, whereby the activity of HPA leads to depletion of free SH- groups in GSH and proteins through the action of 3-hydroxypropionaldehyde, causing an imbalance of the cellular redox status, ultimately resulting in cell death.
Sigma-Aldrich
Propionaldehyde, 97%
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Propionaldehyde, reagent grade, 97%