Merck
  • Evidence for the efficacy of an 8% strontium acetate dentifrice for instant and lasting relief of dentin hypersensitivity.
Evidence for the efficacy of an 8% strontium acetate dentifrice for instant and lasting relief of dentin hypersensitivity.
The Journal of clinical dentistry (2010-07-31)
Teresa Layer, Nathan Hughes
ABSTRACT
An 8% strontium acetate, 1040 ppm sodium fluoride-based dentifrice has been demonstrated in in vitro and in situ studies to provide deep and acid-resistant tubule occlusion. Clinical efficacy studies have shown instant and lasting dentin hypersensitivity relief from this dentifrice, which has recently been launched by GlaxoSmithKline Consumer Healthcare as Sensodyne Rapid Relief.
Sigma-Aldrich
Strontium acetate
Sigma-Aldrich
Strontium acetate, 99.95% trace metals basis