Merck
  • Binding of alkylphenols and alkylated non-phenolics to rainbow trout (Oncorhynchus mykiss) hepatic estrogen receptors.
Binding of alkylphenols and alkylated non-phenolics to rainbow trout (Oncorhynchus mykiss) hepatic estrogen receptors.
Ecotoxicology and environmental safety (2007-05-22)
Knut-Erik Tollefsen, Anja Julie Nilsen
ABSTRACT
Alkylphenols are well-known endocrine disrupters, mediating effects through the estrogen receptor (ER). In the present work, the interaction of alkylphenols and alkylated non-phenolics with hepatic rainbow trout (Oncorhynchus mykiss) estrogen receptors (rtERs) was determined. The role of alkyl chain length and branching, substituent position, number of alkylated groups, and the requirement of a phenolic ring structure was assessed. The results showed that the rtERs bound most alkylphenols, although with 20,000 to 2 million times lower affinity than the endogenous estrogen 17beta-estradiol. Mono-substituted alkylphenols with moderate (C4-C6) and long (C8 and C12) alkyl chain length in the para position exhibited the highest affinity for the rtERs. Substitution with multiple alkyl groups, presence of substituents in the ortho- and meta-position, and lack of a hydroxyl group on the benzene ring reduced the binding affinity. The rtERs resembled the reported binding specificity of the human ER for alkylphenols, although some exceptions were identified.
Sigma-Aldrich
4-tert-Butylbenzoic acid, 99%