Merck
  • [Studies on chemical constituents from roots of Peucedanum praeruptorum II].
[Studies on chemical constituents from roots of Peucedanum praeruptorum II].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-10-26)
Cun Zhang, Yong-Qing Xiao, Massahiko Taniguchi, Kimiye Baba
ABSTRACT
To isolate and identify the chemical constituents from the roots of Peucedantu praeruptorum. The constituents were isolated by column chromatography on silica gel and ODS, and identified by NMR, MS spectroscopic methods. Eight compounds, (-) sclerodin (1), palmitic acid (2), (+)-praeruptorin E (3), bergapten (4), imperatorin (5), 2,6-dimethyl quinoline (6), tetracosanoic acid (7), and daucosterol (8), were isolated and identified. Compounds 1,5-7 were isolated from this plant for the first time.
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)