Merck
  • Transglycosylation of neohesperidin dihydrochalcone by Bacillus stearothermophilus maltogenic amylase.
Transglycosylation of neohesperidin dihydrochalcone by Bacillus stearothermophilus maltogenic amylase.
Journal of agricultural and food chemistry (2000-02-26)
J S Cho, S S Yoo, T K Cheong, M J Kim, Y Kim, K H Park
ABSTRACT
Neohesperidin dihydrochalcone (NHDC), a sweet compound derived from citrus fruits, was modified to a series of its oligosaccharides by transglycosylation activity of Bacillus stearothermophilus maltogenic amylase (BSMA). Maltotriose as a donor was reacted with NHDC as an acceptor to glycosylate for the purpose of increasing the solubility of NHDC. Maltosyl-NHDC was a major transglycosylation product among the several transfer products by TLC analysis. The structure of the major transglycosylation product was determined to be maltosyl-alpha-(1,6)-neohesperidin dihydrochalcone by MALDI-TOF/MS and (1)H and (13)C NMR. Maltosyl-NHDC was 700 times more soluble in water and 7 times less sweet than NHDC.
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥96%, FG
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥95% (HPLC)
Supelco
Neohesperidin dihydrochalcone, analytical standard