Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibition of 4-hydroxyphenylpyruvate dioxygenase by sethoxydim, a potent inhibitor of acetyl-coenzyme A carboxylase.
Inhibition of 4-hydroxyphenylpyruvate dioxygenase by sethoxydim, a potent inhibitor of acetyl-coenzyme A carboxylase.
Bioorganic & medicinal chemistry letters (1999-03-31)
S Lin, D Yang
ABSTRACT
Sethoxydim, a commercially available cyclohexanedione class herbicide by targeting the enzymatic activity of acetyl-coenzyme A carboxylase, has been found to moderately inhibit the activity of 4-hydroxyphenylpyruvate dioxygenase, a key enzyme in the biosynthesis of plastoquinones and tocopherols in plants.
Product Description
Supelco
Sethoxydim, PESTANAL®, analytical standard