Merck
  • Quantitative LC/MS/MS method and pharmacokinetic studies of columbin, an anti-inflammation furanoditerpen isolated from Radix Tinosporae.
Quantitative LC/MS/MS method and pharmacokinetic studies of columbin, an anti-inflammation furanoditerpen isolated from Radix Tinosporae.
Biomedical chromatography : BMC (2007-03-09)
Qirong Shi, Mingjin Liang, Weidong Zhang, Chuan Zhang, Runhui Liu, Yunheng Shen, Huiliang Li, Xiaolin Wang, Xiangwei Wang, Qiongqun Pan, Chunlin Chen
ABSTRACT
Columbin is an important component isolated from Radix Tinosporae. It has been demonstrated to possess many pharmacological activities, including anti-inflammation, antitumor and inhibition of enzyme activity in vivo. The purpose of the present study was to examine in vivo pharmacokinetics and bioavailability of columbin in rats using a high-performance liquid chromatography coupled with tandem mass spectrometry quantitative detection method. The columbin was extracted from rat plasma samples by methyl tert-butyl ether, evaporated and reconstituted in 100 microL methanol prior to analysis. The separation was performed using a Luna reversed-phase analytical column (5 microm, 100 x 2.0 mm) and an SB-C18 guard column (5 microm, 20 x 4.0 mm). The mobile phase was a mixture of methanol and water containing 25 mmoL/L NH(4)Ac (80:20, v/v). The method was validated within the concentration range of 5-5000 ng/mL, and the calibration curves were linear with correlation coefficients (r) >0.999. It was further applied to assess pharmacokinetics and oral bioavailability of columbin after i.v. and oral administration to rats. The oral bioavailability of columbin was only 3.18%, which indicated that columbin had poor absorption or underwent extensive first-pass metabolism.
Sigma-Aldrich
Columbin, ≥95% (LC/MS-ELSD)