Merck
  • Production of polygalacturonases by Aspergillus oryzae in stirred tank and internal- and external-loop airlift reactors.
Production of polygalacturonases by Aspergillus oryzae in stirred tank and internal- and external-loop airlift reactors.
Bioresource technology (2012-09-04)
Roselei Claudete Fontana, Maurício Moura da Silveira
ABSTRACT
The production of endo- and exo-polygalacturonase (PG) by Aspergillus oryzae was assessed in stirred tank reactors (STRs), internal-loop airlift reactors (ILARs) and external-loop airlift reactors (ELARs). For STR production, we compared culture media formulated with either pectin (WBE) or partially hydrolyzed pectin. The highest enzyme activities were obtained in medium that contained 50% pectin in hydrolyzed form (WBE5). PG production in the three reactor types was compared for WBE5 and low salt WBE medium, with additional salts added at 48, 60 and 72h (WBES). The ELARs performed better than the ILARs in WBES medium where the exo-PG was the same concentration as for STRs and the endo-PG was 20% lower. These results indicate that PG production is higher under experimental conditions that result in higher cell growth with minimum pH values less than 3.0.
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid