Merck
  • The catalytic role of glutamate 151 in the leucine aminopeptidase from Aeromonas proteolytica.
The catalytic role of glutamate 151 in the leucine aminopeptidase from Aeromonas proteolytica.
The Journal of biological chemistry (2004-05-13)
Krzysztof P Bzymek, Richard C Holz
ABSTRACT
Glutamate 151 has been proposed to act as the general acid/base during the peptide hydrolysis reaction catalyzed by the co-catalytic metallohydrolase from Aeromonas proteolytica (AAP). However, to date, no direct evidence has been reported for the role of Glu-151 during catalytic turnover by AAP. In order to elucidate the catalytic role of Glu-151, altered AAP enzymes have been prepared in which Glu-151 has been substituted with a glutamine, an alanine, and an aspartate. The Michaelis constant (K(m)) does not change upon substitution to aspartate or glutamine, but the rate of the reaction changes drastically in the following order: glutamate (100% activity), aspartate (0.05%), glutamine (0.004%), and alanine (0%). Examination of the pH dependence of the kinetic constants k(cat) and K(m) revealed a change in the pK(a) of a group that ionizes at pH 4.8 in recombinant leucine aminopeptidase (rAAP) to 4.2 for E151D-AAP. The remaining pK(a) values at 5.2, 7.5, and 9.9 do not change. Proton inventory studies indicate that one proton is transferred in the rate-limiting step of the reaction at pH 10.50 for both rAAP and E151D-AAP, but at pH 6.50 two protons and general solvation effects are responsible for the observed effects in the reaction catalyzed by rAAP and E151D-AAP, respectively. Based on these data, Glu-151 is intrinsically involved in the peptide hydrolysis reaction catalyzed by AAP and can be assigned the role of a general acid and base.
Sigma-Aldrich
Aminopeptidase from Aeromonas proteolytica, lyophilized powder, 50-150 units/mg protein
Sigma-Aldrich
Aminopeptidase His-tagged from Vibrio proteolyticus