Merck
p120-Catenin suppresses NLRP3 inflammasome activation in macrophages.
American journal of physiology. Lung cellular and molecular physiology (2023-03-08)
Paulraj Kanmani, Hoda El-Hossiny Elkafas, Muhammed Ghazal, Richard D Minshall, Guochang Hu
ABSTRACT
Inflammasome activation is of central importance for the process of generation of overwhelming inflammatory response and the pathogenesis of sepsis. The intrinsic molecular mechanism for controlling inflammasome activation is still poorly understood. Here we investigated the role of p120-catenin expression in macrophages in regulating nucleotide-binding oligomerization domain (NOD) and leucine-rich repeat (LRR)- and pyrin domain-containing proteins 3 (NLRP3) inflammasome activation. Depletion of p120-catenin in murine bone marrow-derived macrophages enhanced caspase-1 activation and secretion of active interleukin (IL)-1β in response to ATP stimulation following LPS priming. Coimmunoprecipitation analysis showed that p120-catenin deletion promoted NLRP3 inflammasome activation by accelerating the assembly of the inflammasome complex comprised of NLRP3, apoptosis-associated speck-like protein containing a caspase recruitment domain (ASC), and pro-caspase-1. Depletion of p120-catenin also increased the production of mitochondrial reactive oxygen species. Pharmacological inhibition of mitochondrial reactive oxygen species nearly completely abolished NLRP3 inflammasome activation, caspase-1 activation, and the production of IL-1β in p120-catenin-depleted macrophages. Furthermore, p120-catenin ablation significantly disrupted mitochondrial function, evidenced by decreased mitochondrial membrane potential and lower production of intracellular ATP. In alveolar macrophage-depleted mice challenged with cecal ligation and puncture, pulmonary transplantation of p120-catenin-deficient macrophages dramatically enhanced the accumulation of IL-1β and IL-18 in bronchoalveolar lavage fluid. These results demonstrate that p120-catenin prevents NLRP3 inflammasome activation in macrophages by maintaining mitochondrial homeostasis and reducing the production of mitochondrial reactive oxygen species in response to endotoxin insult. Thus, inhibition of NLRP3 inflammasome activation by stabilization of p120-catenin expression in macrophages may be a novel strategy to prevent an uncontrolled inflammatory response in sepsis.