Merck
  • Detection of invisible biological traces in relation to the physicochemical properties of substrates surfaces in forensic casework.
Detection of invisible biological traces in relation to the physicochemical properties of substrates surfaces in forensic casework.
Scientific reports (2024-06-11)
Mathilde Recipon, Rémy Agniel, Philippe Kunemann, Arnaud Ponche, Franck Carreiras, Francis Hermitte, Johanne Leroy-Dudal, Sylvain Hubac, Olivier Gallet, Sabrina Kellouche
ABSTRACT
Touch DNA, which can be found at crime scenes, consists of invisible biological traces deposited through a person's skin's contact with an object or another person. Many factors influence touch DNA transfer, including the "destination" substrate's surface. The latter's physicochemical characteristics (wettability, roughness, surface energy, etc.) will impact touch DNA deposition and persistence on a substrate. We selected a representative panel of substrates from objects found at crime scenes (glass, polystyrene, tiles, raw wood, etc.) to investigate the impact of these characteristics on touch DNA deposition and detection. These were shown to impact cell deposition, morphology, retention, and subsequent touch DNA genetic analysis. Interestingly, cell-derived fragments found within keratinocyte cells and fingermarks using in vitro touch DNA models could be successfully detected whichever the substrates' physicochemistry by targeting cellular proteins and carbohydrates for two months, indoors and outdoors. However, swabbing and genetic analyses of such mock traces from different substrates produced informative profiles mainly for substrates with the highest surface free energy and therefore the most hydrophilic. The substrates' intrinsic characteristics need to be considered to better understand both the transfer and persistence of biological traces, as well as their detection and collection, which require an appropriate methodology and sampling device to get informative genetic profiles.
Sigma-Aldrich
Anti-Keratin 10 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.