Merck
  • Sitagliptin ameliorates busulfan-induced pulmonary and testicular injury in rats through antioxidant, anti-inflammatory, antifibrotic, and antiapoptotic effects.
Sitagliptin ameliorates busulfan-induced pulmonary and testicular injury in rats through antioxidant, anti-inflammatory, antifibrotic, and antiapoptotic effects.
Scientific reports (2023-06-17)
Eman A Ali, Sara G Tayel, Mona A Abbas
ABSTRACT
Busulfan (BUS) is an anticancer agent with serious adverse effects on various body organs, including the lung and testis. Sitagliptin was proven to have antioxidant, anti-inflammatory, antifibrotic, and antiapoptotic effects. This study aims to evaluate whether sitagliptin, a DPP4I, ameliorates BUS-induced pulmonary and testicular injury in rats. Male Wistar rats were split into control, sitagliptin (10 mg/kg), BUS (30 mg/kg), and sitagliptin + BUS groups. Weight change, lung and testis indices, serum testosterone, sperm parameters, markers of oxidative stress [malondialdehyde (MDA) and reduced glutathione (GSH)], inflammation [tumor necrosis factor-alpha (TNF-α)], and relative expression of sirtuin1 (SIRT1) and forkhead box protein type O1 (FOXO1) genes were estimated. Histopathological examination of lung and testicular tissues was done to detect architectural changes [Hematoxylin & Eosin (H&E)], fibrosis (Masson's trichrome), and apoptosis (caspase-3). Sitagliptin treatment reduced body weight loss, lung index, lung and testis MDA, serum TNF-α and sperm abnormal morphology, and increased testis index, lung and testis GSH, serum testosterone, sperm count, viability and motility. SIRT1/FOXO1 balance was restored. Also, sitagliptin attenuated fibrosis and apoptosis in lung and testicular tissues via reducing collagen deposition and caspase-3 expression. Accordingly, sitagliptin ameliorated BUS-induced pulmonary and testicular damage in rats via attenuating oxidative stress, inflammation, fibrosis, and apoptosis.