Merck
  • Radiation dose-escalation and dose-fractionation modulate the immune microenvironment, cancer stem cells and vasculature in experimental high-grade gliomas.
Radiation dose-escalation and dose-fractionation modulate the immune microenvironment, cancer stem cells and vasculature in experimental high-grade gliomas.
Journal of neurosurgical sciences (2020-10-16)
Matteo Riva, Roxanne Wouters, David Nittner, Jolien Ceuster, Edmond Sterpin, Roberto Giovannoni, Uwe Himmelreich, Willy Gsell, Marc Van Ranst, An Coosemans
ABSTRACT
In the context of high-grade gliomas (HGGs), very little evidence is available concerning the optimal radiotherapy (RT) schedule to be used in radioimmunotherapy combinations. This studied was aimed at shedding new light in this field by analyzing the effects of RT dose escalation and dose fractionation on the tumor microenvironment of experimental HGGs. Neurospheres (NS) CT-2A HGG-bearing C57BL/6 mice were treated with stereotactic RT. For dose-escalation experiments, mice received 2, 4 or 8 Gy as single administrations. For dose-fractionation experiments, mice received 4 Gy as a single fraction or multiple (1.33x3 Gy) fractions. The impact of the RT schedule on murine survival and tumor immunity was evaluated. Modifications of glioma stem cells (GSCs), tumor vasculature and tumor cell replication were also assessed. RT dose-escalation was associated with an improved immune profile, with higher CD8+ T cells and CD8+ T cells / regulatory T cells (Tregs) ratio (p=0.0003 and p=0.0022, respectively) and lower total tumor associated microglia/macrophages (TAMs), M2 TAMs and monocytic myeloid derived suppressor cells (mMDSCs) (p=0.0011, p=0.0024 and p<0.0001, respectively). The progressive increase of RT dosages prolonged survival (p<0.0001) and reduced tumor vasculature (p=0.069), tumor cell proliferation (p<0.0001) and the amount of GSCs (p=0.0132 or lower). Compared to the unfractionated regimen, RT dose-fractionation negatively affected tumor immunity by inducing higher total TAMs, M2 TAMs and mMDSCs (p=0.0051, p=0.0036 and p=0.0436, respectively). Fractionation also induced a shorter survival (p=0.0078), a higher amount of GSCs (p=0.0015 or lower) and a higher degree of tumor cell proliferation (p=0.0003). This study demonstrates that RT dosage and fractionation significantly influence survival, tumor immunity and GSCs in experimental HGGs. These findings should be taken into account when aiming at designing more synergistic and effective radio-immunotherapy combinations.
Sigma-Aldrich
Anti-NES antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution