Merck
  • Determination of aflatoxins B1, G1, B2 and G2 in medicinal herbs by liquid chromatography-tandem mass spectrometry.
Determination of aflatoxins B1, G1, B2 and G2 in medicinal herbs by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2004-09-30)
Meritxell Ventura, Antonio Gómez, Ivan Anaya, Jordi Díaz, Francesc Broto, Montserrat Agut, Lluís Comellas
ABSTRACT
An easy method for the determination of aflatoxins B1, G1, B2 and G2 in Rhammus purshiana by LC coupled to mass spectrometry has been developed. Aflatoxins were extracted with a mixture of methanol and water and then it was purified by solid-phase clean-up using a polymeric sorbent, not described previously, for the determination of these toxins. The eluted extract was injected into the chromatographic system using a reversed-phase C18 short column with an isocratic mobile phase composed of methanol-water (30:70). A single-quadruple mass spectrometry using an electrospray ionization source operating in the positive ion mode was used to detect aflatoxins due to derivatization presenting several disadvantages. Recoveries of the full analytical procedure were 110% for aflatoxin B1, 89% for aflatoxin B2, 81% for aflatoxin G1 and 77% for aflatoxin G2. Detection limit (S/N = 3) was 10 ng and quantification limit (S/N = 10) was 25 ng, calculated as amount in medicinal herb.