Merck
Expedient synthesis of conjugated triynes via alkyne metathesis.
Chemical science (2020-04-27)
Idriss Curbet, Sophie Colombel-Rouen, Romane Manguin, Anthony Clermont, Alexandre Quelhas, Daniel S Müller, Thierry Roisnel, Olivier Baslé, Yann Trolez, Marc Mauduit
ABSTRACT
The first synthesis of conjugated triynes by molybdenum-catalysed alkyne metathesis is reported. Strategic to the success of this approach is the utilization of sterically-hindered diynes that allowed for the site-selective alkyne metathesis to produce the desired conjugated triyne products. The steric hindrance of the alkyne moiety was found to be crucial in preventing the formation of diyne byproducts. This novel synthetic strategy was amenable to self- and cross-metathesis providing straightforward access to the corresponding symmetrical and dissymmetrical triynes with high selectivity.
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%