Merck
  • Age and Body Condition Influence the Post-Prandial Interleukin-1β Response to a High-Starch Meal in Horses.
Age and Body Condition Influence the Post-Prandial Interleukin-1β Response to a High-Starch Meal in Horses.
Animals : an open access journal from MDPI (2021-12-25)
Jessica Suagee-Bedore, Nichola Shost, Christian Miller, Luis Grado, Jeremy Bechelli
ABSTRACT
Older horses and those prone to obesity may be at a higher risk for inflammation than younger and leaner counterparts. Previous research indicated a postprandial elevation in plasma concentrations of interleukin-1β (IL-1β), a pro-inflammatory cytokine, after consuming 1.2 g of non-structural carbohydrates/kilogram of body weight. However, these studies utilized horses of mixed age and body condition. The current study evaluated post-prandial IL-1β concentrations in horses specifically comparing lean to over-conditioned and middle aged to older. Our results suggest that at least two weeks of daily consumption of a high non-structural carbohydrate diet is required to induce a post-prandial increase in IL-1β concentrations in younger and leaner horses. In opposition to this, older and over-conditioned horses experience plasma increased on the first day of feeding and thereafter. Feeding management practices of older and over-conditioned individuals should emphasize lower non-structural carbohydrate intakes and further research should elucidate mechanisms of IL-1β activation.