Merck
  • Iron(III) chloride as an efficient catalyst for stereoselective synthesis of glycosyl azides and a cocatalyst with Cu(0) for the subsequent click chemistry.
Iron(III) chloride as an efficient catalyst for stereoselective synthesis of glycosyl azides and a cocatalyst with Cu(0) for the subsequent click chemistry.
Chemical communications (Cambridge, England) (2011-08-16)
Santosh B Salunke, N Seshu Babu, Chien-Tien Chen
ABSTRACT
A highly efficient and mild method for azido glycosylation of glycosyl β-peracetates to 1,2-trans glycosyl azides was developed by using inexpensive FeCl(3) as the catalyst. In addition, we demonstrated, for the first time, that FeCl(3) in combination with copper powder can promote 1,3-dipolar cycloaddition (click chemistry) of azido glycosides with terminal alkynes. Good to excellent yields were obtained with exclusive formation of a single isomer in both glycosyl azidation and subsequent cycloaddition processes.