Merck
  • 10-Gingerol Targets Lipid Rafts Associated PI3K/Akt Signaling in Radio-Resistant Triple Negative Breast Cancer Cells.
10-Gingerol Targets Lipid Rafts Associated PI3K/Akt Signaling in Radio-Resistant Triple Negative Breast Cancer Cells.
Molecules (Basel, Switzerland) (2020-07-16)
Meran Keshawa Ediriweera, Jeong Yong Moon, Yen Thi-Kim Nguyen, Somi Kim Cho
ABSTRACT
10-gingerol is a major phenolic lipid found in the rhizomes of ginger (Zingiber officinale). Being amphiphilic in nature, phenolic lipids have the ability to incorporate into cell membranes and modulate membrane properties. The purpose of the present study was to evaluate the effects of 10-gingerol on lipid raft/membrane raft modulation in radio-resistant triple negative breast cancer (MDA-MB-231/IR) cells. The effects of 10-gingerol on MDA-MB-231/IR cells' proliferation, clonogenic growth, migration, and invasion were assayed using MTT, colony formation, cell migration, and invasion assays, respectively. Sucrose density gradient centrifugation was used to extract lipid rafts. Western blotting and immunofluorescence were employed to assess the effects of 10-gingerol on lipid raft/membrane raft modulation and lipid rafts-associated PI3K/Akt signaling. Cholesterol measurements were carried out using a commercially available kit. 10-gingerol suppressed the proliferation, migration, invasion, and induced apoptosis through targeting the PI3K/Akt signaling pathway in MDA-MB-231/IR cells. Moreover, 10-gingerol was found to modulate the lipid rafts of MDA-MB-231/IR cells and attenuate the key PI3K/Akt signaling components in lipid rafts. The cholesterol content of the lipid rafts and rafts-resident Akt signaling were also affected by exposure to 10-gingerol. The results of the present study highlight rafts-associated PI3K/Akt signaling as a new target of 10-gingerol in MDA-MB-231/IR cells, thus rationalizing a new rafts-mediated treatment approach for radio-resistant triple negative breast cancer cells.
Sigma-Aldrich
[10]-Gingerol, ≥98% (HPLC)