Merck
  • Development of direct competitive biomimetic immunosorbent assay based on quantum dot label for determination of trichlorfon residues in vegetables.
Development of direct competitive biomimetic immunosorbent assay based on quantum dot label for determination of trichlorfon residues in vegetables.
Food chemistry (2018-02-08)
Qiurui Liu, Mingdi Jiang, Zeliang Ju, Xuguang Qiao, Zhixiang Xu
ABSTRACT
A direct competitive biomimetic immunosorbent assay method based on molecularly imprinted polymer was developed for the determination of trichlorfon. A CdSe/ZnS quantum dot label was used as the marker. The hydrophilic imprinted film was synthesized directly on the surface of a 96-well plate, and characterized by Fourier-transform infrared spectroscopy and thermo-gravimetric analyses. The method exhibited high stability, selectivity, and sensitivity. Under optimal conditions, the limits of detection and sensitivity of the biomimetic immunosorbent assay method were 9.0 μg L-1 and 5.0 mg L-1 (0.1 mg kg-1 and 62.5 mg kg-1 for vegetable sample), respectively. Low cross-reactivity values of 19.2% and 15.6% were obtained for the structural analogues. Spinach and rape samples spiked with trichlorfon were extracted and determined by this method with recoveries ranging from 83.6% to 91.1%. The method was applied for the detection of trichlorfon residues in leek and cucumber samples, and results correlated well with those obtained using GC.
Product Description
Supelco
Trichlorfon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland