Merck
Get up to 22% off for Pi Day until 3/26.Save Now
mTOR: from growth signal integration to cancer, diabetes and ageing.
Nature reviews. Molecular cell biology (2010-12-16)
Roberto Zoncu, Alejo Efeyan, David M Sabatini
ABSTRACT
In all eukaryotes, the target of rapamycin (TOR) signalling pathway couples energy and nutrient abundance to the execution of cell growth and division, owing to the ability of TOR protein kinase to simultaneously sense energy, nutrients and stress and, in metazoans, growth factors. Mammalian TOR complex 1 (mTORC1) and mTORC2 exert their actions by regulating other important kinases, such as S6 kinase (S6K) and Akt. In the past few years, a significant advance in our understanding of the regulation and functions of mTOR has revealed the crucial involvement of this signalling pathway in the onset and progression of diabetes, cancer and ageing.