Merck
  • 1,4-diazepine-2,5-dione ring formation during solid phase synthesis of peptides containing aspartic acid beta-benzyl ester.
1,4-diazepine-2,5-dione ring formation during solid phase synthesis of peptides containing aspartic acid beta-benzyl ester.
Journal of peptide science : an official publication of the European Peptide Society (2007-09-14)
Helga Süli-Vargha, Gitta Schlosser, Janez Ilas
ABSTRACT
The Fmoc-based SPPS of H-Xaa-Asp(OBzl)-Yaa-Gly-NH(2) sequences results in side reactions yielding not only aspartimide peptides and piperidide derivatives, but also 1,4-diazepine-2,5-dione-peptides. Evidence is presented to show that the 1,4-diazepine-2,5-dione derivative is formed from the aspartimide peptide. The rate of this ring transformation depends primarily on the tendency to aspartimide and piperidide formation, which is influenced by the nature of the amino acid following the aspartic acid beta-benzyl ester (Xaa). However the bulkiness of the amino acid side chain preceeding the aspartic acid beta-benzyl ester (Yaa) is also important. Under certain conditions the 1,4-diazepine-2,5-dione peptide derivative may even be formed dominantly, which is a highly undesirable side reaction in peptide synthesis, but which provides a new way for the synthesis of diazepine peptide derivatives with targeted biological or pharmacological activity.
Sigma-Aldrich
L-Aspartic acid β-benzyl ester