Merck
The renaissance of GSK3.
Nature reviews. Molecular cell biology (2001-10-05)
P Cohen, S Frame
ABSTRACT
Glycogen synthase kinase 3 (GSK3) was initially described as a key enzyme involved in glycogen metabolism, but is now known to regulate a diverse array of cell functions. The study of the substrate specificity and regulation of GSK3 activity has been important in the quest for therapeutic intervention.