Merck
  • Meat quality, post-mortem proteolytic enzymes, and myosin heavy chain isoforms of different Thai native cattle muscles.
Meat quality, post-mortem proteolytic enzymes, and myosin heavy chain isoforms of different Thai native cattle muscles.
Animal bioscience (2021-04-27)
Chanporn Chaosap, Panneepa Sivapirunthep, Ronachai Sitthigripong, Piyada Tavitchasri, Sabaiporn Maduae, Tipyaporn Kusee, Jutarat Setakul, Kazeem Adeyemi
ABSTRACT
This study investigated the meat quality characteristics, endogenous proteolytic enzymes, collagen content, and myosin heavy chain (MyHC) isoforms of different muscles of Thai native cattle (TNC). Infraspinatus (IF), Longissimus thoracis (LT), and Supraspinatus (SS) muscles were obtained from two TNC breeds, Kho-Lan (KL, n = 7) and Kho-Isaan (KI, n = 7). The muscle and meat characteristics of TNC breeds and their relationship with MyHC expression were examined. Three MyHC isoforms namely MyHC I, MyHC IIa, and MyHC IIx were detected in the muscles. The KL had higher (p<0.05) MyHC IIx than the KI. The IF muscle had higher (p<0.05) MyHC I compared to other muscles. The LT muscle had the least MyHC I. The LT had higher (p<0.05) MyHC IIx than the IF and SS muscles. The IF presented the least MyHC IIx. The KL had higher (p<0.05) lightness and moisture content and lower crude protein, redness, cooking loss, shear force, and calpastatin than the KI. The glycogen, total collagen, soluble collagen, crude protein, ash contents, and troponin T degradation product of IF and SS were lower (p<0.05) than that of LT. Ether extract in LT was lower (p<0.05) than that of IF and SS. The percentage of MyHC I, MyHC IIa, and MyHC IIx were significantly correlated with muscle and meat characteristics of TNC. These results suggest that the differences in the MyHC isoforms may partly account for the variation in meat quality between breeds and among muscles of TNC.