Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • BDNF and 5-HT: a dynamic duo in age-related neuronal plasticity and neurodegenerative disorders.
BDNF and 5-HT: a dynamic duo in age-related neuronal plasticity and neurodegenerative disorders.
Trends in neurosciences (2004-09-18)
Mark P Mattson, Stuart Maudsley, Bronwen Martin
ABSTRACT
Brain-derived neurotrophic factor (BDNF) and serotonin (5-hydroxytryptamine, 5-HT) are known to regulate synaptic plasticity, neurogenesis and neuronal survival in the adult brain. These two signals co-regulate one another such that 5-HT stimulates the expression of BDNF, and BDNF enhances the growth and survival of 5-HT neurons. Impaired 5-HT and BDNF signaling is central to depression and anxiety disorders, but could also play important roles in the pathogenesis of several age-related disorders, including insulin resistance syndrome, Alzheimer's disease and Huntington's disease. Enhancement of BDNF signaling may be a key mechanism whereby cognitive stimulation, exercise, dietary restriction and antidepressant drugs preserve brain function during aging. Behavioral and pharmacological manipulations that enhance 5-HT and BDNF signaling could help promote healthy brain aging.